Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01312519
 View more versions of this product
Product Code. 10324001
  • 13.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12257
  • Available in a range of socket and thread sizes
  • Minimized need for grease
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
1 Ea.
NS 19/26
GL 25
Duran Glass