Learn More
Lenz Laborglasinstrumente™ Tubes with Socket and Cone
Provides for quick and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Tubes with Socket and Cone are manufactured from durable DURAN™ borosilicate glass.
17.00 € - 20.00 €
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
10589951
|
Lenz Laborglasinstrumente™
01101411 |
Socket Size/Cone Size: NS 14/23 |
17.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
10671892
|
Lenz Laborglasinstrumente™
01101422 |
Socket Size/Cone Size: NS 19/26 |
19.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
11846144
|
Lenz Laborglasinstrumente™
01101444 |
Socket Size/Cone Size: NS 29/32 |
20.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
- Available in a range of socket/cone sizes
- Corrosion-free and resistant to thermal shock
- With conical joint, featurig high taper accuracy and excellent tightness
Specifications
1Pack |