missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ High Temperature red EFTE Pouring ring, GL 32
Screw cap with cap liner sealing and high temperature and chemical resistance.
17.05 € - 25.25 €
Specifications
Color | Red |
---|---|
Description | Pouring ring |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping |
Material | PBT, ETFE, Fiberglass |
Quantity | 10/Pk. |
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | |||||
11722342
|
DWK Life Sciences
292441909 |
GL 32 |
17.05 €
Pack of 10 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11732342
|
DWK Life Sciences
292442802 |
GL 45 |
25.25 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
- replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
- Excellent chemical resistance due to PTFE coating
- Cap liner forms a tight seal with bottle Peroxide catalysed silicone
- Peroxide catalysed silicone
- Suitable for PBT screw caps and tamper-evident screw caps
- Fully compliant with current international food contact regulations
Specifications
Red | |
Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping | |
10/Pk. | |
PTFE-coated Silicone | |
ETFE |
Pouring ring | |
PBT, ETFE, Fiberglass | |
ETFE | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title