Learn More
DWK Life Sciences DURAN™ HPLC Reservoir Bottle, Clear, Conical Base, GL 45
GREENER_CHOICE
The optimized conical design allows the safest and most efficient delivery of mobile phase to HPLC analysers.
Brand:  DWK Life Sciences 218416309
326.87 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15829007
  • 371.00 € / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The DURAN™ reservoir bottles have been specially designed for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents. Reservoirs have a one central cavity for the inlet filter to allow the most efficient delivery of mobile phase to HPLC analysers. A safer and more convenient alternative to flat bottomed glass media bottles, the conical bottom reservoirs allow a greater recovery of solvent, without having to tilt reservoir and risk a solvent spill. The larger reservoirs allow longer, uninterrupted analyses. These are especially useful during a weekend or lengthy chromatographic analysis when there might be a risk of running out of solvents during unattended operation.
  • The optimized conical design allows the safest and most efficient delivery of mobile phase to HPLC analysers
  • Manufactured from the material of choice for solvent bottles: borosilicate 3.3 glass. This material has an exceptional chemical durability offering a very low inorganic ion extractables profile, and the total absence of any organic extractables.
  • DURAN™ is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a Type I, Class A low-expansion borosilicate glass as defined by ASTM-E438-1992
  • DURAN™ glass is classified by the USP / EP / JP pharmacopoeias as a Type I neutral glass
  • Retrace Code, and certificate for manufacturing lot traceability
  • On the two larger sizes, the central cavity can accommodate magnetic stir bars for improved mixing
  • Side vent / drain hole in the base for ultrasonic bath degassing, or water drainage after cleaning
  • GL 45 thread fits standard HPLC connection caps
  • Durable white enamel volume graduations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2000 mL
Supply of solvents to HPLC equipment; Safe transfer of liquid media within a closed and sterile system
1/Pk.
145 mm
Borosilicate Glass (3.3)
Threads, size GL 45
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products