Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable, HA
KPG bearing sleeve is a precisely fitting holder for guiding the KPG agitator shaft.
Brand:  DWK Life Sciences 245004209
34.49 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 10058190
  • 39.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KPG agitators consist of a KPG agitator shaft and a KPG bearing sleeve. KPG bearing parts are manufactured with high accuracy and uniformity. The specified tolerances for nominal diameter and fit are maintained by careful checks. This guarantees the interchangeability of the KPG bearing sleeves and KPG stirrer shafts. Due to the low bearing clearance between the running surfaces, almost absolute sealing and completely quiet running can be achieved with suitable lubrication. KPG agitators are an alternative to magnetic stirrers and are used for reaction vessels that are not accessible from below when very viscous reaction mixtures are to be stirred or a high stirring speed is required.
  • High accuracy and uniformity within defined tolerances for nominal diameter and fit
  • Ground and polished bearing surface
  • Interchangeable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Stirring of liquids (for syntheses); Stirring at high stirring speed; Mounting support
10 mm