Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
969.00 € - 1220.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15884638
|
DWK Life Sciences
286305701 |
Ground Joint |
969.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15804648
|
DWK Life Sciences
286307205 |
Polished |
1220.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
Solid DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
- Solid
- Available in polished and ground version
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding
Specifications
50/Pk. |