Learn More
DWK Life Sciences DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated
Multifunctional glass laboratory bottles with square space saving shape.
106.00 € - 226.00 €
Specifications
For Use With (Application) | Liquid handling reservoir for instruments; Transport and shipping; Sample collection; Storage of media and reagents; Archival storage of samples; Preparation of media and reagents; Culturing vessel |
---|---|
Includes | With screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11716233
|
DWK Life Sciences
218202453 |
100 mL |
106.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11321465
|
DWK Life Sciences
218203655 |
250 mL |
132.10 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11331465
|
DWK Life Sciences
218204454 |
500 mL |
162.02 €
Pack of 10 |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
11341465
|
DWK Life Sciences
218205459 |
1000 mL |
226.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
DURAN™ Square laboratory bottles have a space saving shape that offers the benefit of close and efficient packing. With up to a 20 % saving of space compared to traditional round bottles. Ideal for the shipping and storage of high-demand materials. For reagent supply to instruments where bench space is at a premium. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ square bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Ergonomic handling, high stability and good stackability due to angular shape
- Saves space and costs in autoclaves, drying chambers, ovens, storage or washers
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Liquid handling reservoir for instruments; Transport and shipping; Sample collection; Storage of media and reagents; Archival storage of samples; Preparation of media and reagents; Culturing vessel | |
Borosilicate Glass (3.3) | |
Autoclavable |
With screw cap and pouring ring from PP (blue) | |
10/Pk. | |
Polypropylene |