missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT Product Code.: 10434301
Add to basket

DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT

Product Code. 10434301 Shop All DWK Life Sciences Products
GL 45
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
This item is not returnable. View return policy

Product Code. 10434301

Brand: DWK Life Sciences 292271007

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

PBT screw cap with an open top and high temperature and chemical resistance.

Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.
  • Compatible with GL bottle threads
  • Red PBT offers high temperature and chemical resistance
  • Glass reinforced PBT offers high strength
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Diameter (Metric) Inner Bore Size 34 mm
For Use With (Application) Aseptic reagent delivery and culture sampling using a syringe and needle
Material PBT
Quantity 10/Pk.
Screw Material Polybutylene Terephthalate (PBT) with glass fiber
Thread Size (Metric) GL 45
Autoclavable Autoclavable
Height (Metric) 28 mm
Product Type Open Top Screw Cap
Height (English) 1.1 in.
Closure Material PBT with Glass Fiber
Diameter (Metric) 54 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.