Learn More
DWK Life Sciences DURAN™ Original Laboratory Bottle, Amber, with DIN 168-1 Thread, USP / EP (3.2.1) Light Transmission, Graduated
The gold standard for multifunctional glass laboratory bottles.
216.00 € - 1220.00 €
Specifications
For Use With (Application) | Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel |
---|---|
Includes | With screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11639918
|
DWK Life Sciences
218062951 |
150 mL |
410.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11699918
|
DWK Life Sciences
218065151 |
750 mL |
585.00 €
Pack of 10 |
Estimated Shipment: 01-04-2025
Log in to see stock. |
|||||
11649928
|
DWK Life Sciences
218066953 |
3500 mL |
216.00 €
Each |
In Stock
Log in to see stock. |
|||||
13137256
|
DWK Life Sciences
218062454 |
100 mL |
291.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11689908
|
DWK Life Sciences
218061458 |
25 mL |
265.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11609928
|
DWK Life Sciences
218065451 |
1000 mL |
805.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11659918
|
DWK Life Sciences
218063656 |
250 mL |
483.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11679918
|
DWK Life Sciences
218064455 |
500 mL |
728.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11629928
|
DWK Life Sciences
218066353 |
2000 mL |
1220.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11669928
|
DWK Life Sciences
218068654 |
10,000 mL |
463.00 €
Each |
In Stock
Log in to see stock. |
|||||
11609918
|
DWK Life Sciences
218061758 |
50 mL |
263.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15379849
|
DWK Life Sciences
218060856 |
10 mL |
266.00 €
Pack of 10 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
11659928
|
DWK Life Sciences
218067358 |
5000 mL |
342.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, amber, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The amber bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original amber bottles are supplied with or without iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- UV protection up to approx. 500 nm due to external colouration
- The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
- Very durable and chemically resistant amber colour
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Specifications
Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel | |
With screw cap and pouring ring from PP (blue) | |
Borosilicate Glass (3.3) | |
Autoclavable |