missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only Product Code.: 10051192

DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only

Product Code. 10051192 Shop All DWK Life Sciences Products
25 mL
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20 L
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
750 mL
This item is not returnable. View return policy

Product Code. 10051192

Brand: DWK Life Sciences 1092676

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

DURAN™ Protect Laboratory Bottles are clear, multifunctional borosilicate glass laboratory bottles with a protective safety coating, DIN Threads, and graduations.

  • DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972.
  • The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside.
  • The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
  • A combination of high-performance materials and robust ISO 4796-1 design provides a long working life.
  • The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
  • Permanent Retrace Code on each bottle for manufacturing lot traceability.
  • The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.
  • The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 mL and less).
  • Highly resistance, long service-life coating.
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out.
  • UV protection up to approx. 380 nm due to external coating.
  • Easy-to-read scale with graduation marks to estimate the content.
  • Fired-on, highly durable white ceramic print.
  • Sizes 10 and 25 mL with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 25 mL
Capacity (English) 0.8 oz.
Coating Polyurethane
Color Clear
Description With specially shaped glass edge for improved pouring out (so that an additional plastic pouring ring is not required).
Diameter (English) 1.4 in.
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes Without screw cap and pouring ring
Material Borosilicate Glass (3.3)
Quantity 10 Pk.
Thread Size (Metric) GL 25
Certifications/Compliance ASTM E438, Type I, Class A
Closure Included No
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Narrow
Shape Round
Height (Metric) 70 mm
Product Type Safety-coated Bottle
Height (English) 2.8 in.
Closure Size 25 mm
Diameter (Metric) 36 mm
Show More Show Less
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      GL 45 Protect bottles

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.