Learn More
DWK Life Sciences DURAN™ PTFE Adapter, Standard joint ground 45/40 - GL 45, with EPDM Seal
The DURAN™ PTFE Adapter complements the DURAN™ filtering apparatus, making it possible to filter directly into a DURAN™ pressure plus+ laboratory bottle.
Brand:  DWK Life Sciences 294001207
130.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15393094
  • 148.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • Direct filtration into a DURAN™ pressure plus+ laboratory bottle
  • Reduced risk of contamination as no additional refilling step is required
  • The DURAN™ PTFE Adapter combines the ground joint NS 45/40 of the filtering apparatus with the GL 45 screw thread of the DURAN™ laboratory bottles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
53 mm
1 Ea.
Filtering; As a vacuum is created in the bottle during filtration, use of the DURAN™ pressure plus+ laboratory bottle is recommended
EPDM, PTFE