Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
The screw thread coupling is used to connect two threaded connections.
Brand:  DWK Life Sciences 292260959
 View more versions of this product
Product Code. 10120271
  • 32.65 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
  • For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Connection of glass threads
GL 25
PBT
1 Ea.