Learn More
DWK Life Sciences DURAN™ Slit sieve
For use with paper or membrane filters.
Brand:  DWK Life Sciences 213404404
363.85 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15251689
  • 383.00 € / Pack of 5
Estimated Shipment: 30-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ slotted screen plates serve as support surfaces for the actual filter (e.g. paper or membrane filter), e.g. in the case of buechner funnels.
  • As support for paper or membrane filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
Suitable for coarse filtration without additional filter medium; Filtration
5
5 mm