Learn More
DWK Life Sciences DURAN™ Spherical Joint, cup, polished
For grease-free flexible connections.
42.75 € - 310.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15814668
|
DWK Life Sciences
286325806 |
24 mm |
262.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15884668
|
DWK Life Sciences
286327404 |
28 mm |
310.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15814658
|
DWK Life Sciences
286323008 |
12 mm |
200.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15874658
|
DWK Life Sciences
286324604 |
18 mm |
211.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10355201
|
DWK Life Sciences
286329405 |
48 mm |
42.75 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15864648
|
DWK Life Sciences
286321401 |
8 mm |
197.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
The non-sanded version of the DURAN™ spherical ground joint only seals over the sealing ring. Basically 2 versions of the sealing rings are available: - FEP-coated silicone rings - VITON sealing rings The nominal size is composed of the nominal diameter of the spherical ground joint and, if applicable, the clear width of the pipe diameter. This variant also enables flexible connections, these are in the angle range of 10-15°.
- Flexible connections of up to 15° can be realised depending on the size of the cut (angle ≥ tightness reduced)
- Grease-free working possible
- A tight seal is made possible by the use of an additional sealing ring.
- The nominal size describes the nominal diameter of the spherical ground joint.
- Not self-supporting
- Glass articles for further processing