Learn More
DWK Life Sciences DURAN™ Standard Cone, double, finished
For a rigid, self-supporting connection.
Brand:  DWK Life Sciences 286172809
 View more versions of this product
Product Code. 15844598
  • 304.00 € / Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form D. Their primary purpose is to save material.
  • According to DIN 12249 Form D
  • Ground (finer grinding quality, less roughness depth)
  • Glass article with two cores enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Installation in glass apparatus and devices
13 mm
1.5 mm