Learn More
DWK Life Sciences DURAN™ Standard Cone, finished
For a rigid, self-supporting connection.
3.61 € - 474.00 €
Products 17
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
10706941
View Documents
DWK Life Sciences
286157008
26 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15951531
View Documents
DWK Life Sciences
286153203
13 mm
161.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15864578
View Documents
DWK Life Sciences
286161606
56 mm
265.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15804568
View Documents
DWK Life Sciences
286155007
17 mm
201.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15844558
View Documents
DWK Life Sciences
286151005
6 mm
66.45 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15854558
View Documents
DWK Life Sciences
286152404
11 mm
177.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15804578
View Documents
DWK Life Sciences
286159609
30 mm
442.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
10181601
View Documents
DWK Life Sciences
286160404
90 mm
111.00 €
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15941531
View Documents
DWK Life Sciences
286151802
8 mm
222.00 €
Pack of 50
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
15961531
View Documents
DWK Life Sciences
286156209
22 mm
231.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15834578
View Documents
DWK Life Sciences
286160207
40 mm
117.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15844578
View Documents
DWK Life Sciences
286160601
36 mm
117.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15854578
View Documents
DWK Life Sciences
286161203
46 mm
195.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
10191981
View Documents
DWK Life Sciences
286161409
50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15874578
View Documents
DWK Life Sciences
286161803
65 mm
474.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
10305211
View Documents
DWK Life Sciences
286162002
80 mm
71.30 €
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15834558
View Documents
DWK Life Sciences
286150403
4 mm
67.55 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
Description
Description
The conical ground joint is the most common joint in glass apparatus engineering. Ground DURAN™ cores comply with DIN 12249 Form B in their design and meet higher requirements for surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • According to DIN 12249 Form B
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts.
  • Minimized grease requirement
  • Tight closure due to standard ground joint
  • Glass articles for further processing