Learn More
DWK Life Sciences DURAN™ Standard Socket, diamond-ground
For a rigid, self-supporting connection as a cost-effective alternative.
111.00 € - 182.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15951601
|
DWK Life Sciences
286532806 |
17 mm |
111.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
15824808
|
DWK Life Sciences
286521003 |
15 mm |
115.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15824818
|
DWK Life Sciences
286541202 |
22 mm |
126.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15951611
|
DWK Life Sciences
286558202 |
33 mm |
182.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15804808
|
DWK Life Sciences
286518001 |
13 mm |
121.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15864818
|
DWK Life Sciences
286546003 |
28 mm |
178.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
The conical ground joint of sleeve and core is a rigid, self-supporting joint. The DURAN™ DG sleeves are not ground according to DIN 12249 and represent a cost-effective alternative to the DURAN™ sleeves. In principle, ground joints should be made using ground grease or PTFE sleeves. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
- Diamond cut (coarser cut quality, more roughness depth)
- Cost-effective alternative to DURAN™ sleeves (not according to DIN)
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
50/Pk. |