Learn More
DWK Life Sciences DURAN™ Standard Socket, double, diamond-ground
Designed for a rigid, self-supporting connection with lower requirements.
6.15 € - 254.00 €
Products 3
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
10131411
View Documents
DWK Life Sciences
286573309
34 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15834838
View Documents
DWK Life Sciences
286571205
18 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15854838
View Documents
DWK Life Sciences
286572501
28 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
DURAN™ DG double sleeves are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ sleeves (not according to DIN)
  • Glass article with two sleeves enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing