Learn More
DWK Life Sciences DURAN™ Standard Socket, double, finished
For a rigid, self-supporting connection.
Brand: DWK Life Sciences 286569808
This item is not returnable.
View return policy
Description
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form C. It primarily serves the purpose of saving material.
- According to DIN 12249 Form C
- Glass article with two sleeves enables material savings
- Very low leakage rate
- Minimized grease requirement
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
50/Pk. | |
Installation in glass apparatus and devices | |
18 mm | |
1.8 mm |