Learn More
DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, angled side necks
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
Brand: DWK Life Sciences 241885503
84.23 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
- Tight closure due to standard ground joint
- angled side necks
- Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
- The geometry permits very uniform heating
- Fired-on, highly durable white print

Specifications
131 mm | |
Borosilicate Glass (3.3) | |
1/Pk. | |
Autoclavable |
1000 mL | |
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles | |
Center neck NS 29/32 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.