Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
13.94 € - 275.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
15395817
|
DWK Life Sciences
248416109 |
GL 18, with Cone NS 14/23 |
166.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
13427467
|
DWK Life Sciences
248406202 |
GL 14, with Cone NS 14/23 | N/A | ||||||
13595370
|
DWK Life Sciences
248407207 |
GL 14, with Cone NS 19/26 |
207.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15222009
|
DWK Life Sciences
248409208 |
GL 14, with Cone NS 29/32 |
271.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15305827
|
DWK Life Sciences
248419106 |
GL 18, with Cone NS 29/32 |
261.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15282009
|
DWK Life Sciences
248429201 |
GL 25, with Cone NS 29/32 |
249.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15292009
|
DWK Life Sciences
248449109 |
GL 32, with Cone NS 29/32 |
275.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15242009
|
DWK Life Sciences
248417105 |
GL 18, with Cone NS 19/26 |
175.00 €
Pack of 10 |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
15272009
|
DWK Life Sciences
248428205 |
GL 25, with Cone NS 24/29 |
227.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11639978
|
DWK Life Sciences
248448104 |
GL 32, with Cone NS 24/29 |
256.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
12799696
|
DWK Life Sciences
248418101 |
GL 18, with Cone NS 24/29 |
207.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15656935
|
DWK Life Sciences
248427209 |
GL 25, with Cone NS 19/26 |
207.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
12789696
|
DWK Life Sciences
248408203 |
GL 14, with Cone NS 24/29 |
240.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
- Tight closure due to standard ground joint
- With DIN thread
Specifications
10/Pk. |