Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket
Borosilicate glass reaction vessel with thermostatic jacket
Brand:  Lenz Laborglasinstrumente™ 06010270
 View more versions of this product
Product Code. 10349522
  • 727.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ tubing, cylindrical shape with graduation and laboratory flange LF. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.
Specifications
Specifications
1000 mL
245 mm
Reaction vessel
Flat
Borosilicate Glass, Duran Glass
Thermostatic jacket
Cylindrical