missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Twin-Neck Round Bottom Flask, with standard ground joint, angled side neck

Multi-neck pellets are used for distillation, fraction and reactions of all kinds.

Brand:  DWK Life Sciences 241832604

38.81 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 10597841

  • 49.20 € / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.

  • Tight closure due to standard ground joint
  • angled side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications

Specifications

64 mm
Borosilicate Glass (3.3)
1/Pk.
24/29
Autoclavable
100 mL
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volumen sizes 100 mL (NS 24/29 and NS 29/32), 250 mL (NS 24/29), 500 mL (NS 24/29) and 1000 mL (NS 24/29) are not according to DIN ISO
Non-DIN/ISO size
NS 24/29
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Twin-Neck Round Bottom Flask, with standard ground joint, angled side neck > Center neck NS 24/29, 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.