Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, bent, with 2 standard ground joints, and screw-on plastic hose connection
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
66.20 € - 78.70 €
Specifications
Description Autoclavable
For Use With (Application) Distillation
Fits Tubing ID (Metric) 8.6 mm
Material Borosilicate Glass (3.3), PBT, Polypropylene, Silicone
Quantity 1 Ea.
View More Specs
Products 3
Product Code Brand Thread Size Price Quantity & Availability  
Product Code Brand Thread Size Price Quantity & Availability  
10069940
View Documents
DWK Life Sciences
241302105
NS 14/23
78.70 €
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
10385191
View Documents
DWK Life Sciences
241304603
NS 29/32
71.05 €
Each
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
10633792
View Documents
DWK Life Sciences
241303401
NS 24/29
66.20 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
  • Tight closure due to standard ground joint
  • Bent
  • With two standard ground joint and screw-on plastic hose connection
  • Fired-on, highly durable white print
Specifications
Specifications
Autoclavable
8.6 mm
1 Ea.
Distillation
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone