Learn More
DWK Life Sciences DURAN™ Watch Glass Dish, fused rim
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
59.20 € - 391.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11786592
|
DWK Life Sciences
213212408 |
40 mm |
106.00 €
Pack of 10 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
11796592
|
DWK Life Sciences
213213207 |
50 mm |
116.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11706602
|
DWK Life Sciences
213213404 |
60 mm |
127.00 €
Pack of 10 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
11716602
|
DWK Life Sciences
213214109 |
80 mm |
138.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11726602
|
DWK Life Sciences
213214606 |
100 mm |
157.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11756612
|
DWK Life Sciences
213215208 |
125 mm |
172.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11766612
|
DWK Life Sciences
213215705 |
150 mm |
219.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11716153
|
DWK Life Sciences
213216101 |
200 mm |
391.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10202121
|
DWK Life Sciences
213216607 |
250 mm |
59.20 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
- Reduced risk of injury due to fused rim
Specifications
Autoclavable | |
Borosilicate Glass (3.3) | |
Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass |