Learn More
DWK Life Sciences DURAN™ Weighing Bottle, with ground lid, low form
Weighing glasses are suitable for weighing substances. The matching lid prevents unintentional distribution of the weighing product.
321.00 € - 615.00 €
Specifications
Description Low form
For Use With (Application) Weighing; Storage and transport; Available in low and high forms
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Autoclavable Autoclavable
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11730944
View Documents
DWK Life Sciences
242101304
5 mL
321.00 €
Pack of 10
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
11740944
View Documents
DWK Life Sciences
242102309
15 mL
470.00 €
Pack of 10
In Stock
to see stock.
Add to basket
11700964
View Documents
DWK Life Sciences
242103202
30 mL
615.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11710964
View Documents
DWK Life Sciences
242104104
80 mL
564.00 €
Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
In order to carry out reactions in a controlled manner, it is essential to know the quantities of substances required and to weigh the corresponding substances accurately. A weighing glass can be used for weighing. The associated lid allows the weighing pan to be closed and thus prevents, for example, turbulence of the weighed-in solids during transport. Since even the smallest contamination of the weighing glass can have an effect on the weighing inheritance, the glass in the area of the ground joint and the lid knob are so pronounced that they can be grasped and moved with forceps or tweezers.
  • Close-fitting lid prevents any sample loss during transport after weighing
  • Fired-on, highly durable white ceramic
Specifications
Specifications
Low form
Borosilicate Glass (3.3)
Autoclavable
Weighing; Storage and transport; Available in low and high forms
10/Pk.