Learn More
Macherey-Nagel™ Durasil-25 TLC Plate
Brand: Macherey-Nagel™ 812004
This item is not returnable.
View return policy
Specifications
Durasil-25 | |
60 Å | |
TLC Plate | |
∼500 m2/g | |
20 x 20 cm |
5 to 17 μm | |
25 Pk. | |
0.75 mL/g | |
0.25 mm |