Learn More
Macherey-Nagel™ Durasil-25/UV254 TLC Plate
Brand:  Macherey-Nagel™ 812005
 View more versions of this product
Product Code. 10334601
  • 148.00 € / Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Durasil-25
60 Å
TLC Plate
0.75 mL/g
0.25 mm
5 to 17 μm
50 Pk.
UV254
∼500 m2/g
5 x 10 cm