Learn More
Berkshire International™ DURX™ 670-ROLL
Superior strength and absorbency make Durx™ 670 Rolls an ideal choice for a general purpose wiper.
Brand: Berkshire International™ DR6703438R
This item is not returnable.
View return policy
Description
- Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low.
- Durx™ 670 wipers can be used for cleaning and maintenance in laboratories, aerospace, defense electronics, food processing, computer assembly and semiconductor, circuit board, pharmaceutical, and biomedical device manufacturing.
671 Rolls, General Purpose Wipe, Cleaning Wipe, Durex, Durx wipes
Specifications
Polyester Cellulose | |
Pre-Perforated Sheets | |
34 x 38 cm |