Learn More
Berkshire™ Durx™ 770 Cleanroom wipe
BIOPHARMA
Durx™ 770 is ideal for critical small parts cleaning. Durx™ 770 is enhanced with an ultra-low particle and fiber treatment to significantly reduce particles and fibers compared to Durx™ 670.
Brand:  Berkshire™ DR770070712
Product Code. 12706045
  • 749.00 € / Pack of 3600
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Binder treatment reduces particle and fiber generation. It can be used for cleaning and maintenance in laboratories, aerospace, electronics, food processing, photo processing, and semiconductor, circuit board, pharmaceutical, and biomedical device manufacture.
Specifications
Specifications
Polyester Cellulose
Knife-Cut
ISO Class 5-8
Nonwoven
18 x 18 cm