missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dutasteride, 99%

Product Code. 15608745
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164656-23-9
Molecular Formula C27H30F6N2O2
Molecular Weight (g/mol) 528.53
InChI Key JWJOTENAMICLJG-QWBYCMEYSA-N
Synonym dutasteride, avodart, avolve, duagen, unii-o0j6xjn02i, avodart tn, o0j6xjn02i, dutasteride jan/usan/inn, 5alpha,17beta-n-2,5-bis trifluoromethyl phenyl-3-oxo-4-azaandrost-1-ene-17-carboxamide, dutasteride usan
PubChem CID 6918296
ChEBI CHEBI:521033
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC4=C(C=CC(=C4)C(F)(F)F)C(F)(F)F)CCC5C3(C=CC(=O)N5)C

Specifications

Melting Point 245.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 528.53
Percent Purity 99%
Chemical Name or Material Dutasteride
compliance-icons
Product Identifier
  • Dutasteride
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dutasteride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Dutasteride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.