Learn More
Cole-Parmer™ Dye Concentrate
Determine water flow in closed circuits or open channels with these dyes. Dye Concentrate diffuses rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, flow rates, discharge locations, seepage areas, sea marking and more.
Brand:  Cole-Parmer™ 00298-16
 View more versions of this product
Product Code. 11777947
  • 41.75 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters.
  • Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult
  • Won’t harm the environment—nontoxic and biodegradable
  • Easy-to-use color in rapidly diffusing dye identifies dispersion patterns, flow rates, seepage and more Noncarcinogenic fluorescent red dye is ideal for yellowed or heavily algaed water
  • Brighten murky ponds with blue dye
  • Minimum detectable concentration is 1 ppm
Specifications
Specifications
Dark Red
Squirt-top dispensing
Dye Tracer
8.6 to 8.8
1/Pk.