missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 680 NHS Ester

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Brand:  Thermo Scientific™ 46419

221.16 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 11899370

  • 291.00 € / Pack of 5
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DyLight 680 NHS Ester
Dye
Store desiccated at -20°C.
Active Ester, Succinimidyl Ester, NHS Ester
DyLight Fluors
712
Proteins
5 x 50 μg
DMF (Dimethylformamide)
Amine
DyLight™ 680
DyLight™
692
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DyLight 680 NHS Ester,5 x 50 ug

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : DyLight Reactive Dyes

SDS
Documents

Documents

Certificates
View accessory products
Ofertas especiales

Ofertas especiales

For Research Use Only. Not for use in diagnostic procedures.