Learn More
Thermo Scientific™ DyLight™ 488, Free Acid
Use these non-activated, carboxylic acid forms of DyLight Dyes as controls and calibration standards in fluorescence imaging applications.
Brand: Thermo Scientific™ 62280
219.64 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific™ DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorecence imaging applications.
Thermo Scientific™ DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorecence imaging applications. The non-activated dyes contain the native carboxyl group of the standard dye molecule.
General DyLight Fluorophore Highlights:
- Bright fluorescence — intense emission provides superior sensitivity and requires less conjugate
- Narrow emission spectra — negligible bleed-through between fluorophore channels enables multi-color detection
- Excellent photostability — exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4Pi microscopy)
- Buffer stability — conjugated fluorophores are completely stable at pH 4-9
- Instrument compatibility — excitation and emission spectra correspond with filter sets and laser settings of all popular fluorescence instrumentation
Specifications
DyLight™ 488, Free Acid, 1mg, Solid, DyLight™ 488 Dye, 493/518nm Excitation/Emission Wavelength, <4°C Storage Temperature, Used for Protein Biology, Protein Labeling and Crosslinking, Protein Labeling | |
Free Acid | |
Store at < 4°C. | |
DyLight™ 488 | |
DyLight™ | |
493 |
1 mg | |
DMF (Dimethylformamide) | |
Carboxylic Acid | |
DyLight Fluors | |
518 |
For Research Use Only. Not for use in diagnostic procedures.