missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ mRNA DIRECT™ Purification Kit

Product Code. 10190723
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10190723

Brand: Invitrogen™ 61011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues.

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues. The isolated mRNA is suitable for use in all downstream applications.

Advantages of the Dynabeads mRNA DIRECT Kit:
Fast—15 minute procedure yields pure, intact mRNA
Highly pure mRNA isolation—best choice upstream of cDNA synthesis
Sensitive mRNA isolation—enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)

System overview
The isolation protocol relies on base pairing between the polyA residues at the 3' end of most mRNA, and the oligo (dT)25 residues covalently coupled to the surface of the Dynabeads. Other RNA species lacking a polyA tail will not hybridize to the beads and are readily washed away. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. RNase inhibiting agents in the Lysis/Binding Buffer together with stringent hybridization and washing conditions ensure the isolation of pure, intact mRNA from crude samples rich in RNase, without the use of strong chaotropic agents. The mRNA purification beads specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples (see protocol). 1 mg of Dynabeads Oligo (dT)25 beads (200 μL) binds up to 2 μg of mRNA. A typical mammalian cell contains about 10–30 pg of total RNA, from which 1–5% is mRNA.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• 5 mL Dynabeads oligo (dT)25
• 30 mL Lysis/Binding Buffer
• 60 mL Washing Buffer A
• 30 mL Washing Buffer B
• 15 mL 10 mM Tris-HCl (Elution buffer)

Contains enough reagents for 20 standard isolations. Store at 2 to 8°C.

Isolation Technology Magnetic Bead
Purification Time 15 min.
Sample Type Blood, Cells, FFPE and Fixed Samples, Liquid Samples (e.g. Serum), Plant Samples, RNA, Tissue, Viral Fluids, Yeast
Final Product Type mRNA
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Nuclease Protection Assays, Northern Blotting, Cloning
Product Line DYNAL, Dynabeads, mRNA DIRECT
Quantity 5 mL
Shipping Condition Room Temperature
Target mRNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.