missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Streptavidin for Target Enrichment

Dynabeads Streptavidin for Target Enrichment beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for enrichment of target sequences for library preparations for next-generation sequencing (NGS) workflows.

Brand:  Invitrogen™ 65606D

1362.30 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 17882884

  • 1458.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dynabeads Streptavidin for Target Enrichment beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for enrichment of target sequences for library preparations for next-generation sequencing (NGS) workflows.

Benefits of Dynabeads Streptavidin for Target Enrichment beads include:
Performance—high sensitivity and reproducibility of results
Efficiency—fast binding kinetics and high binding capacity
Accuracy—excellent coverage, which enables high confidence in variant detection
Automation ready—easy automation for high-throughput needs on the KingFisher Purification System

Consistency and reproducibility  
The monolayer of streptavidin leaves most of the biotin-binding sites available for the binding of free biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer help ensure negligible leakage, batch consistency, and reproducibility of results.

High binding capacity 
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads Streptavidin for Target Enrichment typically binds: 
• 950–1,500 pmol free biotin 
• ∼20 μg biotinylated ds-DNA 
• ∼400 pmol biotinylated ss-oligonucleotides

Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophobic beads are particularly well suited for the enrichment of target sequences for library preparations for NGS workflows. Dynabeads Streptavidin for Target Enrichment is a high-quality product that enables robust and reproducible NGS analysis results on any NGS platform.

Dynabeads Streptavidin for Target Enrichment efficiently captures DNA sequences of interest through specific hybridization via immobilization of target-specific biotinylated probes to the streptavidin on the bead surface. The subsequent washing steps utilize magnetic separation to obtain high purity. The starting sample can be nucleic acid extracted from chromatin immunoprecipitated samples (ChIP) or isolated pure nucleic acid.

Automation ready with the KingFisher Purification System
The 1-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems or liquid handlers.

Commercial supply 
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Streptavidin for Target Enrichment beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Magnetic Polystyrene
10 mg/mL
Magnetic beads
KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput Compatible
Streptavidin Magnetic Beads
Ambient Temperature
ISO9001 and ISO13485
Monosized 1 μm (CV <5%)
Brown
Dynabeads Streptavidin for Target Enrichment is supplied in PBS, pH 7.4, with 0.1% BSA, 0.02% NaN3. Store at 2°C to 8°C.

1 μm
Target Sequence Enrichment
Dynabeads™
10 mL
For Research Use Only
Recombinant streptavidin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.