Learn More
ABEBA™ Dynamic Blue Occupational Clogs Ob ESD
Blue dynamic occupational clog with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
Brand:  ABEBA™ 37326/46
 View more versions of this product
Product Code. 17101335
  • 70.95 € / 1 pair
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Breathable, lightweight, comfortable occupational clogs
  • Comply to EN ISO 20347: 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent, blue knitted textile
  • Non foldable heel strap, adjustable
  • Removable active comfort insole
Specifications
Specifications
Blue
46
Unisex
Water Repellent
Knitted Fabric
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC
Clogs
OB SRC ESD