missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx. 0.13g/25 x 25mm

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11301779
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012383.FF
25 x 25 mm N/A
N/A
11450270
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012383.FI
50 x 50 mm N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]

Specifications

7429-91-6
Foil
Dy
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.50
CHEBI:33377
Under Argon
8.55g/cm3
Gray to Silver
99.90%
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Odorless
Gray to silver foil
Dysprosium, Rare Earth Oxides Content
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and dry.

H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III

missing translation for 'einecsNumber' : 231-073-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only