missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx. 0.53g/25 x 25mm

267.33 € - 850.23 €

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391769
SDS View Documents
Thermo Scientific Alfa Aesar
012382.FF
25 x 25 mm
267.33 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11345929
SDS View Documents
Thermo Scientific Alfa Aesar
012382.FI
50 x 50 mm
850.23 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]

Specifications

7429-91-6
Foil
Dy
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.50
CHEBI:33377
Gray to silver foil
Gray to Silver
99.90%
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Odorless
Dysprosium
Videos
Safety and Handling

Safety and Handling

H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III

missing translation for 'einecsNumber' : 231-073-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.