missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11396668

Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11396668
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11396668

Brand: Thermo Scientific Alfa Aesar 013219.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14637-88-8
Molecular Formula C15H21DyO6
Molecular Weight (g/mol) 459.83
MDL Number MFCD00013489
InChI Key VABPDCUJKLETFM-UHFFFAOYSA-N
Synonym dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11
PubChem CID 121233142
IUPAC Name dysprosium;(Z)-4-hydroxypent-3-en-2-one
SMILES [Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 25 g
Formula Weight 459.83 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.