missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 14637-88-8 |
---|---|
Molecular Formula | C15H21DyO6 |
Molecular Weight (g/mol) | 459.83 |
MDL Number | MFCD00013489 |
InChI Key | VABPDCUJKLETFM-UHFFFAOYSA-N |
Synonym | dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 |
PubChem CID | 121233142 |
IUPAC Name | dysprosium;(Z)-4-hydroxypent-3-en-2-one |
SMILES | [Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14637-88-8 | |
459.83 | |
VABPDCUJKLETFM-UHFFFAOYSA-N | |
121233142 | |
[Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21DyO6 | |
MFCD00013489 | |
dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
dysprosium;(Z)-4-hydroxypent-3-en-2-one |
Specifications
14637-88-8 | |
C15H21DyO6 | |
dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
[Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
459.83 | |
459.83 (Anhydrous) | |
Reacton™ | |
Hygroscopic | |
Dysprosium(III) 2,4-pentanedionate hydrate |
Powder | |
MFCD00013489 | |
VABPDCUJKLETFM-UHFFFAOYSA-N | |
dysprosium;(Z)-4-hydroxypent-3-en-2-one | |
121233142 | |
99.9% | |
Odorless | |
(REO) |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only