missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) carbonate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 38245-35-1
Molecular Formula C3H8Dy2O13
Molecular Weight (g/mol) 577.084
MDL Number MFCD00150229
InChI Key BQRZSEZQNZPPPU-UHFFFAOYSA-H
Synonym dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
PubChem CID 25021600
IUPAC Name dysprosium(3+);tricarbonate;tetrahydrate
SMILES C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
015286.09
10 g N/A
N/A
11337108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
015286.18
50 g N/A
N/A
11357108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
015286.30
250 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38245-35-1
577.084
BQRZSEZQNZPPPU-UHFFFAOYSA-H
25021600
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]
C3H8Dy2O13
MFCD00150229
dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
dysprosium(3+);tricarbonate;tetrahydrate

Specifications

38245-35-1
C3H8Dy2O13
dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]
577.084
577.06 (505.03 Anhydrous)
Hygroscopic
Dysprosium(III) carbonate tetrahydrate
Powder
MFCD00150229
BQRZSEZQNZPPPU-UHFFFAOYSA-H
dysprosium(3+);tricarbonate;tetrahydrate
25021600
99.9%
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 225-770-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only