missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Product Code. p-7049267
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11376908

Brand: Thermo Scientific Alfa Aesar 013656.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13569-80-7

Dysprosium(III) fluoride, anhydrous is used for vacuum deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13569-80-7
MDL Number MFCD00016071

Specifications

Boiling Point 2200°C
Melting Point 1360°C
Color White
Physical Form Crystalline Lump
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 219.5
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.