Learn More
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
196.98 € - 1357.00 €
Chemische Identifikatoren
CAS | 15474-63-2 |
---|---|
Molecular Formula | DyI3 |
Molecular Weight (g/mol) | 543.213 |
MDL Number | MFCD00016072 |
InChI Key | RZQFCZYXPRKMTP-UHFFFAOYSA-K |
Synonym | dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrous |
PubChem CID | 84931 |
IUPAC Name | dysprosium(3+);triiodide |
SMILES | [I-].[I-].[I-].[Dy+3] |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11353877
|
Thermo Scientific Alfa Aesar
035693.03 |
1 g |
196.98 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343877
|
Thermo Scientific Alfa Aesar
035693.06 |
5 g |
854.25 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363877
|
Thermo Scientific Alfa Aesar
035693.14 |
25 g | N/A | ||||||
Beschreibung
It is used in high-intensity metal-halide lamps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemische Identifikatoren
15474-63-2 | |
543.213 | |
RZQFCZYXPRKMTP-UHFFFAOYSA-K | |
84931 | |
[I-].[I-].[I-].[Dy+3] |
DyI3 | |
MFCD00016072 | |
dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrous | |
dysprosium(3+);triiodide |
Spezifikation
955°C | |
Green to Yellow | |
DyI3 | |
dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrous | |
RZQFCZYXPRKMTP-UHFFFAOYSA-K | |
dysprosium(3+);triiodide | |
84931 | |
99.99% | |
Hygroscopic | |
Dysprosium(III) iodide, Ultra dry |
15474-63-2 | |
Flakes | |
MFCD00016072 | |
Soluble in water. | |
[I-].[I-].[I-].[Dy+3] | |
543.213 | |
543.21 | |
Ampule | |
(REO) |
Sicherheit und Handhabung
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 239-493-4
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.