missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) oxalate decahydrate, REacton™, 99.9% (REO), Thermo Scientific™

Product Code. 11392538
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24670-07-3
Molecular Formula C6H26Dy2O22
Molecular Weight (g/mol) 775.252
MDL Number MFCD00150482
InChI Key WJYBVSNZIQHDSA-UHFFFAOYSA-N
Synonym scandium oxalate decahydrate
PubChem CID 131876564
IUPAC Name dysprosium;oxalic acid;decahydrate
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Dy].[Dy]

Specifications

Melting Point 40°C (in -H2O)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula Dy2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 769.21 (589.06 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) oxalate decahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'