Learn More
Dysprosium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
Descrizione
A solution of 2,6-diformyl-4-methylphenol (0.15mmol) and benzoylhydrazide (0.30 mmol) in MeOH (10 ml) was allowed to react with the dysprosium(III) perchlorate hexahydrate (0.3 mmol) in 10 ml MeOH In observation of slow magnetic relaxation in triple-stranded lanthanide helicates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica
Specifica
Physical Form | Liquid |
Quantity | 2 g |
UN Number | UN3211 |
Solubility Information | Insoluble in water. |
Formula Weight | 568.94 (460.85 Anhydrous) |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Chemical Name or Material | Dysprosium(III) perchlorate hexahydrate |
Sicurezza e movimentazione


- Dysprosium perchlorate, 50% in water
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Perchloric acid, dysprosium(3+) salt, hexahydrate
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.