missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
A solution of 2,6-diformyl-4-methylphenol (0.15mmol) and benzoylhydrazide (0.30 mmol) in MeOH (10 ml) was allowed to react with the dysprosium(III) perchlorate hexahydrate (0.3 mmol) in 10 ml MeOH In observation of slow magnetic relaxation in triple-stranded lanthanide helicates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Quantity | 10 g |
UN Number | UN3211 |
Formula Weight | 568.94 (460.85 Anhydrous) |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Chemical Name or Material | Dysprosium(III) perchlorate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'