missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester, 98%, Thermo Scientific™

Product Code. 11325465
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185427-48-9
Molecular Formula C18H34B2O4
Molecular Weight (g/mol) 336.09
MDL Number MFCD03093898,MFCD03093898
InChI Key SACOFNGKSPZAFQ-UHFFFAOYSA-N
Synonym e-1-hexene-1,2-diboronic acid bis pinacol ester, 1e-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-ene, z-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-hexene, 4,4,5,5-tetramethyl-2-1z-2-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane
PubChem CID 6026893
IUPAC Name 4,4,5,5-tetramethyl-2-[(Z)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hex-1-en-2-yl]-1,3,2-dioxaborolane
SMILES CCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1

Specifications

Density 0.954
Boiling Point 102°C to 104°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.456
Quantity 5 g
Beilstein 7768251
Formula Weight 336.09
Percent Purity 98%
Chemical Name or Material (E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'