missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For use with the 3M™ One-Touch™ Dispenser
- Refill bag contains 500 pairs of 3M™ Classic™ Earplugs
- Earplug dispenser serves as a reminder to wear hearing protection and make earplugs readily available in noisy areas for full time users and visitors
- Moisture-resistant material helps maintain a consistent expansion time, even in humid environments
- Textured surface helps keep earplugs in place

Specifications
Specifications
Color | Yellow |
Packaging Type | Top-Up Bag |
Pairs per Package | 500 Pairs/Bag |
Noise Reduction Rating (NRR) | 28 dB |
Product Type | Ear Plugs |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ E-A-R™ Classic™ Earplugs PD-01-009 >
Spot an opportunity for improvement?Share a Content Correction