missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ E-A-R™ Classic™ Earplugs Refill Bottle
Click to view available options
For Use With (Application):
E-A-R™ Classic™
Description
- Soft, energy-absorbing foam
- Textured surface helps keep plugs in place
- Moisture-resistant to maintain a consistent expansion time, even in humid environments
- Dispenser serves as a reminder to wear hearing protection and make earplugs readily available
Specifications
Specifications
Color | Yellow |
For Use With (Application) | E-A-R™ Classic™ |
Pairs per Package | 500 Pairs |
Product Type | Ear Plug Refill Pack |
Dielectric | No |
Material | PVC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'